Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Apolipoprotein F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101430
Description
Antibody detects endogenous levels of total APOF.
The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.
Specifications
Apolipoprotein F | |
Polyclonal | |
Unconjugated | |
APOF | |
AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
319 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13790 | |
APOF | |
A synthesized peptide derived from human APOF(Accession Q13790), corresponding to amino acid residues L293-I326. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction