Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein H/ApoH Antibody (SR1979), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Apolipoprotein H/ApoH |
---|---|
Clone | SR1979 |
Dilution | Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Apolipoprotein H/ApoH | |
Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
A synthesized peptide derived from human Apolipoprotein H/ApoH (Uniprot #: P02749) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1979 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cholesterol Metabolism, Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
350 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title