Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein H/ApoH Mouse anti-Human, Non-human Primate, Clone: H219, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3182861000UG
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human, Non-human Primate samples. It is validated for Western Blot, ELISASpecifications
| Apolipoprotein H/ApoH | |
| Monoclonal | |
| 0.5 mg/ml | |
| 0.2 um filtered PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Non-human Primate | |
| Purified |
| Western Blot, ELISA | |
| H219 | |
| Unconjugated | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Purified human Apolipoprotein H/ApoH | |
| 1000 μg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 350 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction