Learn More
Invitrogen™ Apolipoprotein L1 Recombinant Rabbit Monoclonal Antibody (1A8N2)

Description
Immunogen sequence: RPRVTEPISA ESGEQVERVN EPSILEMSRG VKLTDVAPVS FFLVLDVVYL VYESKHLHEG AKSETAEELK KVAQELEEKL NILNNNYKIL QADQEL.
Specifications
Specifications
| Antigen | Apolipoprotein L1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 1A8N2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | APOL1 |
| Gene Accession No. | O14791 |
| Gene Alias | APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.