Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582641 Supplier Invitrogen™ Supplier No. PA582641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNFLSLAGNT YQLTRGIGKD IRALRRARAN LQSVPHASAS RPRVTEPISA ESGEQVERVN EPSILEMSRG VKLTDVAPVS FFLVLDVVYL VYESKHLHEG AKSETAEELK KVAQELEEKL NILNN Highest antigen sequence indentity to the following orthologs: Rat - 39%, Mouse - 43%.

APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOL1
Gene Accession No. O14791
Gene Alias APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705
Gene Symbols APOL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOL1. Recombinant protein control fragment (Product #RP-100508).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.