Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein L1 Recombinant Rabbit Monoclonal Antibody (32H8)
GREENER_CHOICE

Catalog No. FEMA550120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550120 Supplier Invitrogen™ Supplier No. MA550120
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Apolipoprotein L1 Recombinant Monoclonal Antibody for Flow, ELISA

APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L1
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 32H8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APOL1
Gene Accession No. O14791
Gene Alias APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705
Gene Symbols APOL1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human APOL1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.