Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Apolipoprotein L2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519016
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| Apolipoprotein L2 | |
| Polyclonal | |
| Unconjugated | |
| APOL2 | |
| Apol2; ApoL-2; APOL3; Apol8; APOL-II; apolipoprotein L 8; apolipoprotein L, 2; Apolipoprotein L2; Apolipoprotein L-2; apolipoprotein L-II; OTTHUMP00000028936 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23780 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BQE5 | |
| APOL2 | |
| Synthetic peptide sequence (TKIHEMLQPGQD) corresponding to the C-terminus amino acids of APOL2 (aa 325-337). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction