Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein L4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562049 Supplier Invitrogen™ Supplier No. PA562049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSTDQLEALR DILRDITPNV LSFALDFDEA TKMIANDVHT LRRSKATVGR PLIAWRYVPI NVVETLRTRG APTRIVRKVA RNL Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOL4
Gene Accession No. Q9BPW4
Gene Alias APOL4; apolipoprotein L, 4; Apolipoprotein L4; apolipoprotein L-IV; APOLIV; APOL-IV; OTTHUMP00000028937
Gene Symbols APOL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Apolipoprotein L4. Recombinant protein control fragment (Product #RP-101146).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.