Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein L6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583068 Supplier Invitrogen™ Supplier No. PA583068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDNQAERESE AGVGLQRDED DAPLCEDVEL QDGDLSPEEK IFLREFPRLK EDLKGNIDKL RALADDIDKT HKKFTKAN Highest antigen sequence indentity to the following orthologs: Rat - 32%, Mouse - 38%.

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOL6
Gene Accession No. Q9BWW8
Gene Alias APOL6; apolipoprotein L, 6; Apolipoprotein L6; apolipoprotein L-VI; APOLVI; APOL-VI; DKFZp667M075; FLJ38562; FLJ90164; MGC57495; UNQ3095/PRO21341
Gene Symbols APOL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOL6. Recombinant protein control fragment (Product #RP-95374).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.