Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein L6 Polyclonal Antibody

Supplier: Thermo Scientific PA5113622
Description
Specifications
| Apolipoprotein L6 | |
| Polyclonal | |
| Unconjugated | |
| APOL6 | |
| APOL6; apolipoprotein L, 6; Apolipoprotein L6; apolipoprotein L-VI; APOLVI; APOL-VI; DKFZp667M075; FLJ38562; FLJ90164; MGC57495; UNQ3095/PRO21341 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80830 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BWW8 | |
| APOL6 | |
| Synthetic peptide directed towards the middle region of human APOL6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction