Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein L6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518979
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518979 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518979 Supplier Invitrogen™ Supplier No. PA518979
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene APOL6
Gene Accession No. Q9BWW8
Gene Alias APOL6; apolipoprotein L, 6; Apolipoprotein L6; apolipoprotein L-VI; APOLVI; APOL-VI; DKFZp667M075; FLJ38562; FLJ90164; MGC57495; UNQ3095/PRO21341
Gene Symbols APOL6
Host Species Goat
Immunogen Synthetic peptide sequence (QRDEDDAPLCED) corresponding to the internal amino acids of APOL6 (aa 16-28).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80830
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.