Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein M/ApoM Mouse anti-Human, Clone: M5, Novus Biologicals™

Mouse Monoclonal Antibody
$312.52 - $1068.12
Specifications
| Antigen | Apolipoprotein M/ApoM |
|---|---|
| Clone | M5 |
| Concentration | 0.5 mg/ml |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
Apolipoprotein M/ApoM Monoclonal antibody specifically detects Apolipoprotein M/ApoM in Human samples. It is validated for Western Blot, ELISASpecifications
| Apolipoprotein M/ApoM | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered PBS | |
| 55937 | |
| IgG1 | |
| Protein G purified |
| M5 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| apolipoprotein M, apo-M, ApoM, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a | |
| Recombinant human Apolipoprotein M/ApoM | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title