Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein M Monoclonal Antibody (8F12C6B8)
GREENER_CHOICE

Catalog No. PIMA515322
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515322 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515322 Supplier Invitrogen™ Supplier No. MA515322
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15322 targets ApoM in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15322 immunogen is purified recombinant fragment of human ApoM expressed in E. Coli. MA5-15322 detects ApoM which has a predicted molecular weight of approximately 21kDa.

The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein M
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8F12C6B8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene APOM
Gene Accession No. O95445
Gene Alias 1190010O19Rik; alternative name: G3a, NG20; Apo M; Apolipoprotein M; APOM; apo-M; G3A; HSPC336; MGC22400; NG20; NG20 like protein; NG20-like protein; protein G3a; Protein Px
Gene Symbols APOM
Host Species Mouse
Immunogen Purified recombinant fragment of human ApoM expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55937
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.