Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Apolipoprotein M Recombinant Rabbit Monoclonal Antibody (4G4N1)
Apolipoprotein M Recombinant Rabbit Monoclonal Antibody (ARC1210)
Supplier: Invitrogen™ MA535659
Description
Apolipoprotein M Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.
Specifications
Apolipoprotein M | |
Recombinant Monoclonal | |
0.47 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95445, Q9Z1R3 | |
APOM | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ApoM (O95445). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
4G4N1 | |
Unconjugated | |
APOM | |
1190010O19Rik; alternative name: G3a, NG20; Apo M; Apolipoprotein M; APOM; apo-M; G3A; HSPC336; MGC22400; NG20; NG20 like protein; NG20-like protein; protein G3a; Protein Px | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55937, 55938 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction