Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein O Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12502R

Description
This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16.Specifications
Apolipoprotein O | |
Polyclonal | |
Unconjugated | |
APOO | |
0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; Fam121b; family with sequence similarity 121B; MGC4825; Mic23; Mic26; MICOS complex subunit MIC23; MICOS complex subunit Mic26; My025; MYO25; Protein FAM121B; UNQ1866/PRO4302 | |
Rabbit | |
Protein A | |
RUO | |
79135 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BUR5 | |
APOO | |
KLH conjugated synthetic peptide derived from human ApoO/Apolipoprotein O. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction