Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoM/Apolipoprotein M Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982289
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2289 Supplier Bioss Supplier No. BS12501R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.

Specifications

Antigen ApoM/Apolipoprotein M
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene APOM
Gene Accession No. O95445
Gene Alias 1190010O19Rik; alternative name: G3a, NG20; Apo M; Apolipoprotein M; APOM; Apo-M; G3A; HSPC336; MGC22400; NG20; NG20 like protein; NG20-like protein; Protein G3a; Protein Px
Gene Symbols APOM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ApoM/Apolipoprotein M.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55937
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.