Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOOL Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305931MFV610
Description
APOOL Polyclonal antibody specifically detects APOOL in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
APOOL | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
AAIR8193, apolipoprotein O-like, chromosome X open reading frame 33, CXorf33, FAM121A, family with sequence similarity 121A, MGC129748, Protein FAM121A, UNQ8193 | |
Produced in rabbits immunized with E. coli-derived Human APOOL fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
139322 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction