Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOOL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599585 Supplier Invitrogen™ Supplier No. PA599585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ApoOL.

This gene encodes a protein which contains an apolipoprotein O superfamily domain. This domain is found on proteins in circulating lipoprotein complexes. [provided by RefSeq, Sep 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOOL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APOOL
Gene Accession No. Q6UXV4, Q78IK4
Gene Alias 6720473G16Rik; 9430083G14Rik; AAIR8193; apolipoprotein O like; apolipoprotein O-like; Apool; CXorf33; E130106L15Rik; FAM121A; family with sequence similarity 121A; MIC27; MICOS complex subunit MIC27; Protein FAM121A; RGD1549725; UNQ8193; UNQ8193/PRO23204
Gene Symbols APOOL
Host Species Rabbit
Immunogen A synthesized peptide derived from human APOOL(Accession Q6UXV4), corresponding to amino acid residues S228-S268.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139322, 317191, 68117
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.