Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOPT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113262
Description
APOPT1 Polyclonal Antibody for Western Blot, ELISA
APOPT1 plays a role in the regulation of apoptosis. Mediates mitochondria-induced cell death in vascular smooth muscle cells through the release of cytochrome c from mitochondria, followed by the activation of the caspase cascade.
Specifications
| APOPT1 | |
| Polyclonal | |
| Unconjugated | |
| APOPT1 | |
| 1700081D05Rik; 2810002N01Rik; APOP; Apop1; APOP-1; APOPT1; apoptogenic 1, mitochondrial; apoptogenic protein 1, mitochondrial; apoptogenic, mitochondrial 1; C14orf153; C21H14orf153; Coa8; Cytochrome c oxidase assembly factor 8; cytochrome c oxidase assembly factor 8; apoptogenic protein 1, mitochondrial; hypothetical protein LOC84334; LOW QUALITY PROTEIN: apoptogenic protein 1, mitochondrial; UPF0671 protein C14orf153; UPF0671 protein C14orf153 homolog | |
| Rabbit | |
| Protein G | |
| RUO | |
| 68020, 84334 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96IL0, Q9CQW7 | |
| APOPT1 | |
| Recombinant Human Apoptogenic protein 1, mitochondrial protein (40-150AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction