Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOPT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570949 Supplier Invitrogen™ Supplier No. PA570949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 82%; Horse: 82%; Human: 100%.

APOPT1 plays a role in the regulation of apoptosis. Mediates mitochondria-induced cell death in vascular smooth muscle cells through the release of cytochrome c from mitochondria, followed by the activation of the caspase cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOPT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene APOPT1
Gene Accession No. Q96IL0
Gene Alias 1700081D05Rik; 2810002N01Rik; APOP; Apop1; APOP-1; APOPT1; apoptogenic 1, mitochondrial; apoptogenic protein 1, mitochondrial; apoptogenic, mitochondrial 1; C14orf153; C21H14orf153; Coa8; Cytochrome c oxidase assembly factor 8; cytochrome c oxidase assembly factor 8; apoptogenic protein 1, mitochondrial; hypothetical protein LOC84334; LOW QUALITY PROTEIN: apoptogenic protein 1, mitochondrial; UPF0671 protein C14orf153; UPF0671 protein C14orf153 homolog
Gene Symbols APOPT1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human APOPT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84334
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.