Learn More
Invitrogen™ Apoptosis-Enhancing Nuclease Polyclonal Antibody

Description
Immunogen sequence: SLTIPNAKDV LRKRHKRRSR QHQRFMARKA LLQEQGLLSM PPEPGSSPLP TPFGAATATE AASSGKQCLR AGSGSAP Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 62%.
Specifications
Specifications
| Antigen | Apoptosis-Enhancing Nuclease |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AEN |
| Gene Accession No. | Q8WTP8 |
| Gene Alias | 2700083B06Rik; AEN; aen {ECO:0000250; apoptosis enhancing nuclease; apoptosis-enhancing nuclease; apoptosis-enhancing nuclease {ECO:0000250; interferon stimulated exonuclease gene 20kDa-like 1; interferon stimulated exonuclease gene 20-like 1; interferon-stimulated 20 kDa exonuclease-like 1; interferon-stimulated 20 kDa exonuclease-like 1 {ECO:0000312; ISG20L1; pp12744; RGD:1305051}; RGD1305051; SBBI58; UniProtKB:Q8WTP8} |
| Gene Symbols | AEN |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.