Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apoptosis enhancing nuclease Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094020 20 μL
NB094019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094020 Supplier Novus Biologicals Supplier No. NBP30344620UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apoptosis enhancing nuclease Polyclonal specifically detects Apoptosis enhancing nuclease in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Apoptosis enhancing nuclease
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias apoptosis enhancing nuclease, EC 3.1, EC 3.1.13.1, FLJ12484, FLJ12562, interferon stimulated exonuclease gene 20kDa-like 1, Interferon-stimulated 20 kDa exonuclease-like 1, ISG20L1apoptosis-enhancing nuclease, pp12744
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human Apoptosis enhancing nuclease (NP_073604.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64782
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.