Learn More
Description
- Cells undergoing apotosis are irreversibly labeled with a cell permeable, FITC-conjugated pan-caspase inhibitor (ApoStat)
- Assayed by flow cytometry for detection of the bound reagent
- Increased fluorescence is an indicator of caspases activity within cells
- Sample Type: Cultured cells
Specifications
Specifications
| For Use With (Application) | Identification and quantification of caspase activity in apoptotic cells by flow cytometry |
| Content And Storage | Store the unopened product at -20 to -70°C |
| Conjugate | FITC |
| Quantity | 100 Tests |
| Product Type | ApoStat Apoptosis Detection Kit |
| Product Line | R&D Systems |
For research use only. Not for use in diagnostic procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
