Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP+1 Rabbit anti-Human, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF850SP
This item is not returnable.
View return policy
Description
APP+1 Polyclonal antibody AF850SP specifically detects APP+1 in Human samples. It is validated for Western BlottingSpecifications
APP+1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Rabbit | |
Affinity Purified | |
RUO | |
351 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
APP | |
KLH-coupled human APP+1 synthetic peptide, CMRMGRGRTSSKELA | |
25 μg | |
Primary | |
Detects human APP+1 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction