Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP Antibody (6E10) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP2625660.025MG
Description
APP Monoclonal specifically detects APP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
APP | |
Monoclonal | |
Unconjugated | |
PBS | |
APP | |
Beta Amyloid signal peptide 1-16. | |
351 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy | |
60000000000 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Electron Microscopy 1:10 - 1:500 | |
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
Rabbit | |
0.025 mg | |
This antibody binds to amino acid residues 1-16 of beta amyloid. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction