Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306205C
Description
APP Polyclonal specifically detects APP in Human samples. It is validated for ELISA.Specifications
| APP | |
| Polyclonal | |
| ELISA | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| Produced in rabbits immunized with purified, recombinant Human APP (P05067-1; Asp672-Val711). | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 351 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction