Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Mouse anti-Human, Clone: OTI2B10, lyophilized, TrueMAB™
SDP

Catalog No. 501660012
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-0012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-0012 Supplier OriGene Supplier No. CF500973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa.

Specifications

Antigen APP
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene APP
Gene Accession No. P05067
Gene Alias AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2, preA4
Gene Symbols APP
Host Species Mouse
Immunogen Full length human recombit protein of human APP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.