Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP Mouse anti-Mouse, Human, Clone: 2B3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN85225UG
Description
Anti-APP, clone 2B3 is a highly specific mouse monoclonal antibody that targets Amyloid beta A4 protein and has been tested for use in ELISA, Immunocytochemistry, Inhibiton Assay, and Western Blotting.
Specifications
APP | |
Monoclonal | |
Please refer to lot specific datasheet. | |
APP,A4,AD1 | |
A linear peptide corresponding to 15 amino acids from the C-terminal region of human Amyloid-beta A4 protein. | |
25 μg | |
Neuroscience | |
NP_000475 | |
Mouse, Human | |
Purified |
ELISA, Immunocytochemistry, Inhibition Assays, Western Blot | |
2B3 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 2B3 specifically detects Amyloid-beta A4 protein in human and murine neurons. It targets an epitope within 15 amino acids including the beta-secretase cleavage site. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction