Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABN1642MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1642MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1642MI Supplier MilliporeSigma Supplier No. ABN1642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-APP, AEP-cleaved (585N) is a highly specific rabbit polyclonal antibody that targets the N-terminal fragment of AEP-cleaved APP at residue 585 (585N) and has been tested in Immunohistochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols APP,A4,AD1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal end sequence of AEP-cleaved APP 585N fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_958817
Test Specificity This antiserum detects the N-terminal fragment of AEP-cleaved APP at residue 585 (585N), but not the C-terminal fragment (585C) or uncleaved APP. The 585 numbering is based on isoform APP695/PreA4 695 (UniProt P05067-4) sequence. Equivalent site is also present in isoforms APP770 (Asn660; P05067-1), L-APP677 (Asn567; P05067-3), L-APP696 (Asn586; P05067-5), APP714 (Asn604; P05067-6), L-APP733 (Asn623; P05067-7), APP751/PreA4 751 (Asn641; P05067-8), L-APP752 (Asn642; P05067-9), APP639 (Asn529; P05067-10), and isoform 11 (Asn636; P05067-11). Cleavage site is not present in APP305 (P05067-2).
Target Species Mouse, Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.