Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP585C Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $437.74
Specifications
Antigen | APP585C |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN1641-25UL
|
MilliporeSigma
ABN164125UL |
25 μL |
Each for $192.35
|
|
|||||
ABN1641MI
|
MilliporeSigma
ABN1641 |
100 μL |
Each for $437.74
|
|
|||||
Description
Anti-APP585C is a highly specific rabbit polyclonal antibody that targets Amyloid beta A4 protein and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
APP585C | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
APP,A4,AD | |
IgG | |
Unpurified | |
This polyclonal antibody detects amyloid precursor protein in human Alzheimer's disese brain. It targets an epitope within 9 amino acids in the vicintiy of APP585 in the C-terminal region. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Serum | |
RUO | |
Mouse, Human | |
A linear peptide corresponding to 9 amino acids from the C-terminal region of human Amyloid beta A4 protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title