Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APPD Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987502
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7502 Supplier Bioss Supplier No. BS6793R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.

Specifications

Antigen APPD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLEKHF1
Gene Accession No. Q96S99
Gene Alias 1810013P09Rik; AI037048; Apoptosis-inducing protein; apoptosis-inducing protein D; APPD; LAPF; Lysosome-associated apoptosis-inducing protein containing PH and FYVE domains; PH and FYVE domain-containing protein 1; PH domain-containing family F member 1; phafin 1; PHAFIN1; Phafin-1; pleckstrin homology and FYVE domain containing 1; pleckstrin homology domain containing, family F (with FYVE domain) member 1; Pleckstrin homology domain-containing family F member 1; Plekhf1; SGP-1; ZFYVE15; Zinc finger FYVE domain-containing protein 15
Gene Symbols PLEKHF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APPD/ Apoptosis-inducing protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79156
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.