Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215433
Description
APPL Polyclonal specifically detects APPL in Human, Mouse samples. It is validated for Western Blot.Specifications
APPL | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
Rabbit | |
80 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot | |
0.56mg/mL | |
Western Blot 1:500-1:3000 | |
Q9UKG1 | |
APPL1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human APPL1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
26060 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction