Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPL Antibody (OTI4B11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271782F
Description
APPL Monoclonal specifically detects APPL in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
APPL | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
APPL1 | |
Full length recombinant protein of human APPL (NP_036228) produced in HEK293T cells. | |
0.1 mL | |
26060.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4B11 | |
Immunohistochemistry | |
Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction