Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518643
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518643 Supplier Invitrogen™ Supplier No. PA518643
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APPL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene APPL1
Gene Accession No. Q9UKG1
Gene Alias 2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AI585782; AKT2 interactor; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; DIP13 alpha; DIP13A; DIP13alpha; dip13-alpha; Kiaa1428; MODY14; RGD1309388; signaling adaptor protein DIP13alpha
Gene Symbols APPL1
Host Species Goat
Immunogen Synthetic peptide sequence (NRYSRLSKKRENDK) corresponding to the internal amino acids of APPL1 (aa 145-159).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26060
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.