Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Supplier:  Applied Biosystems™ 4303441

Encompass_Preferred

 View more versions of this product

Catalog No. 43-034-41


Only null left
Add to Cart

Description

Description

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mM
Liquid
6 x 1 mL
Standard PCR
GeneAmp dNTP Blend, 2.5 mM (6 x 1 mL)

Store in a constant temperature freezer at –20°C.
GeneAmp™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.