Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Fast Clear Reaction Plates with Barcode
Click to view available options
Quantity:
20 plates
500 plates
Description
- Optimized for performance with Applied Biosystems instruments
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
- Great mechanical stability and flatness for complete workflow needs
- Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability
- Ideal choice for use with automation/robotics and multi-instrument experiments (Like thermal cycler, real-time thermal cycler, genetic analyzer)
- Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
- Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity
- Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
Specifications
| Barcode | Includes Barcode |
| Certifications/Compliance | Certified DNA/RNase Free |
| Content And Storage | Store at room temperature. |
| Packaging | 20 Plates |
| Product Line | MicroAmp |
| Color | Clear, Optical |
| Product Type | Microplate |
| For Use With (Equipment) | Thermal Cycler, Genetic Analyzer, DNA Analyzer |
| Skirt Style | Semi |
| Material | Polycarbonate, Polypropylene |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction