Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Reagents Starter Kit

Catalog No. 4352405
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-524-05 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-524-05 Supplier Applied Biosystems™ Supplier No. 4352405
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG, Eukaryotic 18S rRNA TaqMan™ Assay (with two unlabeled PCR primers and a labeled TaqMan™ probe), plus a sample of human Raji cDNA

TaqMan™ PCR Starter Kit is designed to detect and quantify Eukaryotic 18S rRNA sequence in human Raji cDNA using the TaqMan™ 5' nuclease assay with the TaqMan™ PCR Master Mix.

Gene-specific, ready to use TaqMan™ Gene Expression Assays and Universal Master Mix provide you with a fast, simple way for measuring changes in gene expression.

  • Simple procedure gives you quick results
  • Hot-Start DNA Polymerase enzyme system minimizes nonspecific product amplification for reliable results
  • Used With: 7500 Real-Time PCR System with Dell™ Notebook, 7500 Real-Time PCR System with Dell™ Tower, 7900HT Fast Real-Time PCR System with 384-well Block Module, 7900HT Fast Real-Time PCR System with 384-well Block Module and Automation Accessory, 7900HT Fast Real-Time PCR System with Standard 96-well Block Module, 7900HT System Automation Accessory Upgrade, 7900HT System Standard 384-well Block Upgrade Kit, 7900HT System Standard 96-well Block Upgrade Kit, ABI PRISM™ 7000 Sequence Detection System (Notebook)

Ready-to-Use Gene Expression Solution:
  • The TaqMan™ Reagents Starter Kit contains a sample of TaqMan™ Universal PCR Master Mix, and TaqMan™ Gene-Expression Assay (Eukaryotic 18S rRNA), plus a sample of human Raji cDNA - everything you need for 18S gene-expression analysis
  • Simply combine these reagents according to the instructions to generate reproducible and quantitative gene-expression data

A Complete, Integrated Gene-Expression System:
  • Eliminate time-consuming master mix formulation and assay optimization with the TaqMan™ Gene-Expression Assays and the TaqMan™ Universal PCR Master Mix, by using universal cycling conditions and pre-optimized reagents

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG, Eukaryotic 18S rRNA TaqMan™ Assay (with two unlabeled PCR primers and a labeled TaqMan™ probe), plus a sample of human Raji cDNA.

Store the PCR mix at 2° to 8°C.
Store the rest of the kit contents at -15° to -25°C.
Detection Method Primer-probe
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7500 System, AB 7900 HT 384-Well System
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type PCR Reagents
Quantity 1 kit
Shipping Condition Dry Ice
Target 18S rRNA
Target Specificity Target-Specific
For Use With (Application) Gene Expression
Hot Start Built-In Hot Start
Sample Type DNA (Genomic), cDNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.