Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Universal Master Mix II, no UNG

Catalog No. p-4919406
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-400-43 1 x 1 mL
44-400-40 1 x 5 mL
44-400-47 2 x 5 mL
44-400-48 5 x 5 mL
44-400-49 10 x 5 mL
44-400-41 1 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 44-400-43 Supplier Applied Biosystems™ Supplier No. 4440043
Only null left
Add to Cart
Edge
Add to Cart

Requires

Primers, probes, template, water

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR).

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR). Use TaqMan Universal Master Mix II to amplify complementary DNA (cDNA) and DNA targets for a variety of applications, including quantitation and genotyping.

This TaqMan Universal Master Mix II is supplied as a 2X concentrate and contains:
• AmpliTaq Gold DNA Polymerase, UP (Ultra Pure)
• dNTPs (with dUTP)
• ROX Passive Reference
• Optimized buffer components

Note: This TaqMan Universal Master Mix II does not contain uracil-N-glycoslyase (UNG).

TaqMan Universal Master Mix II has been optimized for use with primers and TaqMan probes that have been designed according to Thermo Fisher Scientific guidelines. The master mix can be used with custom TaqMan assays available from our custom assay service or with pre-optimized assays, such as:
• TaqMan gene expression assays
• TaqMan microRNA assays
• TaqMan drug metabolism genotyping assays
• TaqMan SNP genotyping Assays

For RNA quantitation experiments, TaqMan Universal Master Mix II is used in the second step of a two-step reverse transcription/polymerase chain reaction (RT-PCR) protocol. The cDNA template used with the master mix can be generated in a reverse transcription reaction using cDNA synthesis kits available from Thermo Fisher Scientific.

Alternative product: Try TaqMan Fast Advanced Master Mix (Cat. No. 4444963), our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal Master Mix II and added additional capabilities for your gene expression analysis.

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type Universal Master Mix II
Purity or Quality Grade UP (Ultra Pure)
Quantity 1 x 1 mL
Shipping Condition Wet Ice
For Use With (Application) Gene Expression, Genotyping, miRNA analysis
Fidelity (vs. Taq) 2 X
Sample Type DNA (Genomic), cDNA
Volume 1 mL
Show More Show Less
I want to purchase TaqMan Array Human MicroRNA A Cards v2.0 (Cat. No. 4398965). What other reagents do I need to purchase in addition to these cards?

In order to perform reverse transcription on these cards, you will need the following:

  • TaqMan MicroRNA Reverse Transcription Kit (Cat. No. 4366596)
  • Megaplex RT Primers pools (for human Type A cards Cat. No. 4399966, for Type B cards Cat. No. 4444281)
You can proceed with or without a preamplification step which depends on the total RNA amount. Total RNA amount of 1‐1000 ng supports reverse transcription reaction with preamplification, whereas 350‐1000 ng of total RNA supports a reaction without preamplification. In case you are performing preamplification you would also need the TaqMan PreAmp Master Mix (Cat. No. 4391128) and the Megaplex PreAmp Primers, Human Pool A v2.1 (Cat. No. 4399233). You will also need the Master Mix for the qPCR reaction. It is not included in the card kit. The recommended Master Mix is: TaqMan Fast Advanced Master Mix – run mode Fast (Cat. No. 4444556) For further information, please see the following link.

What is the benchtop stability of the TaqMan miRNA Assay mixed with TaqMan Universal Master Mix II (Cat. No. 4440043)?

24 hours.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.