Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqPath™ ProAmp™ Master Mix

Catalog No. p-7074248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 10 mL
1 x 50 mL
2 x 10 mL
2 x 50 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30865 1 x 1 mL
A30866 1 x 10 mL
A30871 2 x 10 mL
A30867 1 x 50 mL
A30872 2 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A30865 Supplier Applied Biosystems™ Supplier No. A30865
Only null left
Add to Cart
Add to Cart

TaqPath ProAmp Master Mix is for high-throughput genotyping and copy number variation protocols requiring uncompromising reliability and accuracy, even in the presence of inhibitors commonly found in clinical samples.

TaqPath™ ProAmp™ Master Mix

qPCR master mix for multiplex SNP genotyping and purified DNA–based CNV analysis

TaqPath™ ProAmp™ Master Mix is a qPCR master mix designed for high-throughput multiplex SNP genotyping from purified genomic DNA or crude lysates and for copy number variation (CNV) analysis using purified genomic DNA. The formulation delivers sensitive, reproducible amplification performance even in the presence of PCR inhibitors commonly found in biological samples such as blood, buccal swabs, and card punches.

For SNP genotyping applications, TaqPath ProAmp Master Mix supports accurate and reproducible results using either purified genomic DNA or crude lysate sample inputs. For copy number variation analysis, purified genomic DNA is required.

Manufactured under a robust ISO 13485 quality management system, TaqPath ProAmp Master Mix is designed to deliver consistent lot-to-lot performance for high-throughput and regulated research environments.

Designed for multiplex SNP genotyping

A TaqMan™ SNP Genotyping Assay targets a single SNP locus (a specific genomic position) and includes two PCR primers and two allele-specific probes. Each probe hybridizes to the same genomic region but differs by one nucleotide, allowing detection of one of the two possible alleles at that locus. This design enables allelic discrimination within a single qPCR reaction.

TaqPath™ ProAmp™ Master Mix supports singleplex and multiplex SNP genotyping, including 2-SNP multiplex configurations (two SNP loci per reaction; four allele-specific probes total). This multiplexing approach increases data output per sample while maintaining confident allelic discrimination and assay performance.

Six SNP loci per reaction multiplexing requires a custom TaqPath ProAmp formulation without ROX.

Benefits of TaqPath™ ProAmp™ Master Mix

  • Exceptional data quality—high specificity, reproducibility, and dynamic range* for SNP genotyping and CNV analysis
  • Inhibitor tolerance—robust performance with samples derived from blood, buccal swabs, and card punches
  • Optimized for SNP genotyping from crude lysates—supports accurate SNP genotyping without DNA purification
  • 72-hour pre-PCR benchtop stability with purified DNA—ideal for automated and high-throughput workflows, enabled by the proprietary Dual-Lock™ Taq DNA hot-start mechanism
  • Multiplex flexibility—supports singleplex through four-target SNP detection per reaction
  • Manufacturing consistency—produced in an ISO 13485-certified facility
  • Compliance Package available—includes commonly requested quality and regulatory documentation

Accurate SNP genotyping from purified DNA or crude lysates

TaqPath™ ProAmp™ Master Mix delivers accurate and reproducible SNP genotyping results from either purified genomic DNA or crude lysate inputs. When combined with the DNA Extract All™ Reagents Kit, the master mix supports robust singleplex and 2-locus multiplex SNP genotyping directly from crude lysates prepared from blood and buccal swabs.

Internal testing demonstrated 100% concordance between SNP genotype calls obtained from crude lysates and purified genomic DNA using selected 2-SNP multiplex assay configurations, confirming reliable allelic discrimination without DNA purification.

Accurate SNP genotyping from crude lysates with improved performance versus alternative master mixes

In internal evaluations using TaqMan™ SNP Genotyping Assays and crude blood lysates, TaqPath™ ProAmp™ Master Mix demonstrated improved SNP genotype call accuracy and clearer allelic discrimination compared to selected alternative qPCR master mixes. The formulation produced tight genotype cluster separation and consistent SNP locus calls across replicates, supporting confident genotyping performance even in the presence of inhibitors commonly found in biological samples.

High-accuracy copy number variation (CNV) analysis

In addition to SNP genotyping, TaqPath™ ProAmp™ Master Mix supports accurate copy number variation (CNV) analysis when used with purified genomic DNA. Crude lysate sample inputs are not recommended for CNV analysis.

The formulation enables reliable copy number determination across multiple gene targets, including CYP2D6 intron 2 and CYP2D6 intron 6, with accurate quantification in samples containing one to four copies. Internal comparisons demonstrated improved copy number precision and reproducibility compared to selected alternative qPCR master mixes when purified genomic DNA was used as the input.

Part of a streamlined Sample-to-Genotype workflow

TaqPath™ ProAmp™ Master Mix integrates seamlessly into a complete Sample-to-Genotype workflow, including:

  • DNA Extract All™ Reagents Kit for rapid preparation of crude lysates for SNP genotyping
  • TaqMan™ Predesigned SNP Genotyping Assays or TaqMan™ DME Genotyping Assays
  • Purified genomic DNA for copy number variation analysis
  • QuantStudio™ Real-Time PCR Instruments
  • Diomni™ Design and Analysis Software for multiplex genotyping data analysis

This workflow enables faster turnaround and reduced hands-on time for SNP genotyping, while supporting accurate CNV analysis using purified DNA when required.

General purpose reagents manufactured under cGMP

TaqPath™ master mixes are general purpose reagents labeled For Laboratory Use and are manufactured in an ISO 13485-certified and FDA-registered facility that adheres to current Good Manufacturing Practice (cGMP) principles.

Compliance Package

Frequently requested compliance documents are available, including:

  • Statement of Quality describing adherence to cGMP
  • Lot-specific Certificate of Analysis (COA)
  • Product Stability Statement
  • Supplier Evaluation Support documentation
  • Completed Self Assessment
  • ISO certificates by manufacturing site
  • BSE/TSE Statement
  • Change Control and Notification Registration

For EU REACH compliance-related inquiries, email LSGChemicalCompliance@thermofisher.com.

Not finding the document you need? Technical Support is available to help.

* Dynamic range depends on assay design, template concentration, and master mix formulation; individual results may vary.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C in the dark.
Detection Method Primer-probe
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, QuantStudio 12k Flex, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 7 Flex, ViiA 7 System, ViiA 7 Dx, QuantStudio Dx, StepOnePlus System, StepOne System
Multiplex Capability 4-plex
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqPath
Product Type qPCR Master Mix
Quantity 1 x 1 mL
Shipping Condition Wet Ice
Sufficient For 200 Reactions
Target Specificity cDNA or gDNA only
For Use With (Application) Genotyping
Thermostability 72 hr., Assembled Reaction
No. of Reactions 200 Reactions
Volume 1 x 1 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) Very High
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.