Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568745
Description
This target displays homology in the following species: Human: 100%; Pig: 85%; Rabbit: 93%; Rat: 79%.
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
Specifications
APR3 | |
Polyclonal | |
Unconjugated | |
ATRAID | |
0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; all-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein APR-3; apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; C11H2orf28; C17H2orf28; C2orf28; H1E6 protein; HSPC013; LOW QUALITY PROTEIN: all-trans retinoic acid-induced differentiation factor; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51374 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UW56 | |
ATRAID | |
synthetic peptide directed towards the middle region of human APR3 (aa 127-176). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction