Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100702 Supplier Invitrogen™ Supplier No. PA5100702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total APR3.

Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APR3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATRAID
Gene Accession No. Q6PGD0, Q6UW56
Gene Alias 0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; all-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein APR-3; apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; C11H2orf28; C17H2orf28; C2orf28; H1E6 protein; HSPC013; LOW QUALITY PROTEIN: all-trans retinoic acid-induced differentiation factor; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240
Gene Symbols ATRAID
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATRAID(Accession Q6UW56), corresponding to amino acid residues L31-S81.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298841, 381629, 51374
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.