Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aprataxin Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100534B
Description
Aprataxin Polyclonal antibody specifically detects Aprataxin in Human samples. It is validated for Western BlotSpecifications
Aprataxin | |
Polyclonal | |
Western Blot | |
AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Aprataxin using the numbering given in SwissProt Entry Q7Z2E3 (GeneID 54840). | |
0.1 mL | |
Cancer, DNA Repair, Tumor Suppressors | |
54840 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction