Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprataxin Monoclonal Antibody (2H6)
GREENER_CHOICE

Catalog No. PIMA522070
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522070 Supplier Thermo Scientific Supplier No. MA522070
Add to Cart
Add to Cart

Aprataxin Monoclonal Antibody (2H6)

Aprataxin Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Aprataxin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H6
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene APTX
Gene Accession No. Q7Z2E3
Gene Alias 2410016G21Rik; AA388047; AOA; AOA1; Aprataxin; aprataxin isoform a; Aptx; APTX protein; AXA1; EAOH; EOAHA; FHA-HIT; Forkhead-associated domain histidine triad-like protein; forkhead-associated domain histidine-triad like protein
Gene Symbols APTX
Host Species Mouse
Immunogen APTX (NP_778241.1, 69 a.a. ~ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54840
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.