Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aprataxin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541687 Supplier Invitrogen™ Supplier No. PA541687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VIEMVQEAGR VTVRDGMPEL LKLPLRCHEC QQLLPSIPQL KEHLRKHWTQ Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of the histidine triad (HIT) superfamily, some of which have nucleotide-binding and diadenosine polyphosphate hydrolase activities. The encoded protein may play a role in single-stranded DNA repair. Mutations in this gene have been associated with ataxia-ocular apraxia. Multiple transcript variants encoding distinct isoforms have been identified for this gene, however, the full length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aprataxin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene APTX
Gene Accession No. Q7Z2E3
Gene Alias 2410016G21Rik; AA388047; AOA; AOA1; Aprataxin; aprataxin isoform a; Aptx; APTX protein; AXA1; EAOH; EOAHA; FHA-HIT; Forkhead-associated domain histidine triad-like protein; forkhead-associated domain histidine-triad like protein
Gene Symbols APTX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human APTX (aa 307-356).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54840
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.