Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aprataxin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584675 Supplier Invitrogen™ Supplier No. PA584675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HMVNELYPYI VEFEEEAKNP GLETHRKRKR SGNSDSIERD AAQEAEAGTG LEPGSNSGQC SVPLKKGKDA PIKKESLGHW SQGLKISMQD PKMQVYKD Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 69%.

This gene encodes a member of the histidine triad (HIT) superfamily, some of which have nucleotide-binding and diadenosine polyphosphate hydrolase activities. The encoded protein may play a role in single-stranded DNA repair. Mutations in this gene have been associated with ataxia-ocular apraxia. Multiple transcript variants encoding distinct isoforms have been identified for this gene, however, the full length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aprataxin
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APTX
Gene Accession No. Q7Z2E3
Gene Alias 2410016G21Rik; AA388047; AOA; AOA1; Aprataxin; aprataxin isoform a; Aptx; APTX protein; AXA1; EAOH; EOAHA; FHA-HIT; Forkhead-associated domain histidine triad-like protein; forkhead-associated domain histidine-triad like protein
Gene Symbols APTX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APTX. Recombinant protein control fragment (Product #RP-103579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.