Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Aprepitant
GREENER_CHOICESDP

Catalog No. 648650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 64-865-0 Supplier Tocris Bioscience™ Supplier No. 6486/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent long-acting hNK-1 antagonist

Aprepitant is a potent long-acting hNK-1 antagonist, (IC50 = 0.09 nM and Kd = 19 pM). Inhibits induced foot-tapping in gerbils. CNS penetrant and orally bioavailable. Antiemetic.

Specifications

Chemical Name or Material 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
CAS 170729-80-3
Quantity 50 mg
Molecular Formula C23H21F7N4O3
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.