Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APRG1/C3orf35 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982292
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2292 Supplier Bioss Supplier No. BS12505R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Note: This locus is considered non-coding by other groups due to a lack of experimental support for the protein, but NCBI annotates the protein because it meets minimal RefSeq quality criteria for representation. The coding status remains uncertain. [18 Nov 2014].

Specifications

Antigen APRG1/C3orf35
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C3orf35
Gene Accession No. Q8IVJ8
Gene Alias AP20 region protein 1; AP20 region protein1; APRG1; APRG1 tumor suppressor candidate; C3orf35; chromosome 3 open reading frame 35
Gene Symbols C3orf35
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APRG1/C3orf35.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339883
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.