Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548735
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548735 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548735 Supplier Invitrogen™ Supplier No. PA548735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APRG1 Polyclonal Antibody for Western Blot

C3orf35 (Chromosome 3 Open Reading Frame 35) is a Protein Coding gene. Diseases associated with C3orf35 include Pulmonary Hemosiderosis and Muir-Torre Syndrome. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen APRG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene C3orf35
Gene Accession No. Q8IVJ8
Gene Alias AP20 region protein 1; AP20 region protein1; APRG1; APRG1 tumor suppressor candidate; C3orf35; chromosome 3 open reading frame 35
Gene Symbols C3orf35
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 59-88 amino acids from the Central region of human APRG1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339883
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.