Learn More
APRIL Monoclonal Antibody (Sacha-1), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA121169
Description
MA1-21169 detects APRIL in Human and Mouse samples. MA1-21169 has been successfully used in FACS and immunocytochemistry procedures. MA1-21169 immunogen is a recombit fusion protein of resideus 105-250 of human APRIL fused to residues 16-108 of human headless ACRP30
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
Specifications
APRIL | |
Monoclonal | |
1 mg/mL | |
TNFSF13 | |
APRIL, CD256, TALL2, TRDL-1, UNQ383/PRO715, TNFSF13 | |
Rat | |
Size-exclusion - Gel filtration | |
RUO | |
69583, 8741 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Sacha-1 | |
Unconjugated | |
O75888, Q9D777 | |
TNFSF13 | |
Recombit fusion protein of resideus 105-250 of human APRIL fused to residues 16-108 of human headless ACRP30 | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.