Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APRIN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19012125UL
Description
APRIN Polyclonal specifically detects APRIN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
APRIN | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Androgen-induced proliferation inhibitorKIAA0979androgen induced inhibitor of proliferation, Androgen-induced prostate proliferative shutoff-associated protein AS3, androgen-induced shutoff 3, AS3APRIN, CG008, FLJ23236, PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae), sister chromatid cohesion protein PDS5 homolog B | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PDS5B | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ESPESSAIESTQSTPQKGRGRPSKTPSPSQPKKNVRVGRSKQAATKENDSSEEVDVFQGSSPVDDIPQEETEEEEVSTVNVRRRSAKRER | |
25 μL | |
Apoptosis, Cell Cycle and Replication, DNA Repair | |
23047 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction